siRNA Details
| Gene Symbol: BC052688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028430.1 | 4 | 1153 | |||
| GenBank mRNA | AK167253.1 | - | 4 | 1150 | ||
| BC052688.1 | - | 4 | 1108 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC052688| Gene Symbol: BC052688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028430.1 | 4 | 1153 | |||
| GenBank mRNA | AK167253.1 | - | 4 | 1150 | ||
| BC052688.1 | - | 4 | 1108 | |||