siRNA Details
| Gene Symbol: Zfp85os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027969.1 | 3 | 753 | |||
| GenBank mRNA | AK015755.1 | - | 3 | 421 | ||
| AK019608.1 | - | 3 | 470 | |||
| AK134579.1 | - | 3 | 754 | |||
| BC145678.1 | - | 1 | 14 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp85os| Gene Symbol: Zfp85os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027969.1 | 3 | 753 | |||
| GenBank mRNA | AK015755.1 | - | 3 | 421 | ||
| AK019608.1 | - | 3 | 470 | |||
| AK134579.1 | - | 3 | 754 | |||
| BC145678.1 | - | 1 | 14 | |||