siRNA Details
| Gene Symbol: AA387883 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030678.1 | 2 | 1115 | |||
| GenBank mRNA | AK043996.1 | - | 2 | 1116 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AA387883| Gene Symbol: AA387883 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030678.1 | 2 | 1115 | |||
| GenBank mRNA | AK043996.1 | - | 2 | 1116 | ||