siRNA Details
| Gene Symbol: Gm13315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028497.1 | 1 | 320 | |||
| GenBank mRNA | CN704203.1 | - | 1 | 321 | ||
| CN704214.1 | - | 1 | 321 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13315| Gene Symbol: Gm13315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028497.1 | 1 | 320 | |||
| GenBank mRNA | CN704203.1 | - | 1 | 321 | ||
| CN704214.1 | - | 1 | 321 | |||