siRNA Details
| Gene Symbol: BC018473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003364.1 | 4 | 1328 | |||
| GenBank mRNA | AF118558.1 | - | 1 | 419 | ||
| AK156340.1 | - | 4 | 1160 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC018473| Gene Symbol: BC018473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003364.1 | 4 | 1328 | |||
| GenBank mRNA | AF118558.1 | - | 1 | 419 | ||
| AK156340.1 | - | 4 | 1160 | |||