siRNA Details
| Gene Symbol: BC018473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003364.1 | 4 | 3231 | |||
| GenBank mRNA | AF118558.1 | - | 1 | 2320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC018473| Gene Symbol: BC018473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003364.1 | 4 | 3231 | |||
| GenBank mRNA | AF118558.1 | - | 1 | 2320 | ||