siRNA Details
| Gene Symbol: 2610035D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015556.1 | 3 | 1270 | |||
| GenBank mRNA | AK011682.1 | - | 3 | 813 | ||
| AK031204.1 | - | 3 | 1271 | |||
| BC024713.1 | - | 2 | 1208 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610035D17Rik| Gene Symbol: 2610035D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015556.1 | 3 | 1270 | |||
| GenBank mRNA | AK011682.1 | - | 3 | 813 | ||
| AK031204.1 | - | 3 | 1271 | |||
| BC024713.1 | - | 2 | 1208 | |||