siRNA Details
Gene Symbol: 0610038B21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028125.1 | 1 | 1203 | |||
GenBank mRNA | AK002788.1 | - | 1 | 1202 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 0610038B21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028125.1 | 1 | 1203 | |||
GenBank mRNA | AK002788.1 | - | 1 | 1202 |