siRNA Details
| Gene Symbol: 0610038B21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028125.1 | 1 | 1345 | |||
| GenBank mRNA | AK002788.1 | - | 1 | 1344 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

0610038B21Rik| Gene Symbol: 0610038B21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028125.1 | 1 | 1345 | |||
| GenBank mRNA | AK002788.1 | - | 1 | 1344 | ||