siRNA Details
| Gene Symbol: 2610028E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015560.1 | 6 | 2768 | |||
| GenBank mRNA | AK031953.1 | - | 6 | 2768 | ||
| AK082619.1 | - | 6 | 2693 | |||
| AK083757.1 | - | 7 | 3182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610028E06Rik| Gene Symbol: 2610028E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015560.1 | 6 | 2768 | |||
| GenBank mRNA | AK031953.1 | - | 6 | 2768 | ||
| AK082619.1 | - | 6 | 2693 | |||
| AK083757.1 | - | 7 | 3182 | |||