siRNA Details
| Gene Symbol: Thap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028429.1 | 1 | 358 | |||
| GenBank mRNA | AK039179.1 | - | 1 | 337 | ||
| AK080650.1 | - | 1 | 334 | |||
| BB621588.1 | - | 1 | 359 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thap6| Gene Symbol: Thap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028429.1 | 1 | 358 | |||
| GenBank mRNA | AK039179.1 | - | 1 | 337 | ||
| AK080650.1 | - | 1 | 334 | |||
| BB621588.1 | - | 1 | 359 | |||