siRNA Details
| Gene Symbol: Thap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028429.1 | 3, 4 | 947 | |||
| GenBank mRNA | AK015967.1 | - | 4, 5 | 1184 | ||
| AK039179.1 | - | 2, 3 | 823 | |||
| AK080650.1 | - | 2, 3 | 822 | |||
| BC089377.1 | - | 3, 4 | 496 | |||

Thap6
Genomic Map