siRNA Details
| Gene Symbol: Thap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028429.1 | 3 | 824 | |||
| GenBank mRNA | AK015967.1 | - | 4 | 1061 | ||
| AK039179.1 | - | 2 | 700 | |||
| AK080650.1 | - | 2 | 699 | |||
| BC089377.1 | - | 3 | 373 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thap6| Gene Symbol: Thap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028429.1 | 3 | 824 | |||
| GenBank mRNA | AK015967.1 | - | 4 | 1061 | ||
| AK039179.1 | - | 2 | 700 | |||
| AK080650.1 | - | 2 | 699 | |||
| BC089377.1 | - | 3 | 373 | |||