siRNA Details
| Gene Symbol: 4933406F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015568.2 | 2 | 436 | |||
| GenBank mRNA | AK016686.1 | - | 2 | 438 | ||
| AK019801.1 | - | 323 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933406F09Rik| Gene Symbol: 4933406F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015568.2 | 2 | 436 | |||
| GenBank mRNA | AK016686.1 | - | 2 | 438 | ||
| AK019801.1 | - | 323 | ||||