siRNA Details
| Gene Symbol: Cfap53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029424.1 | 1, 2 | 164 | |||
| GenBank mRNA | AK016821.1 | - | 1, 2 | 143 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap53| Gene Symbol: Cfap53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029424.1 | 1, 2 | 164 | |||
| GenBank mRNA | AK016821.1 | - | 1, 2 | 143 | ||