siRNA Details
| Gene Symbol: Cfap53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029424.1 | 2, 3 | 392 | |||
| GenBank mRNA | AK016821.1 | - | 2, 3 | 371 | ||
| BC125572.1 | - | 1, 2 | 50 | 40130087 | ||
| BC125574.1 | - | 1, 2 | 50 | 40130089 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap53| Gene Symbol: Cfap53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029424.1 | 2, 3 | 392 | |||
| GenBank mRNA | AK016821.1 | - | 2, 3 | 371 | ||
| BC125572.1 | - | 1, 2 | 50 | 40130087 | ||
| BC125574.1 | - | 1, 2 | 50 | 40130089 | ||