siRNA Details
| Gene Symbol: 1810014B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015572.2 | 3 | 1103 | |||
| GenBank mRNA | AK007490.1 | - | 3 | 1104 | ||
| AK012122.1 | - | 3 | 1149 | |||
| AK049515.1 | - | 2 | 1121 | |||
| AK090016.1 | - | 3 | 1147 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810014B01Rik| Gene Symbol: 1810014B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015572.2 | 3 | 1103 | |||
| GenBank mRNA | AK007490.1 | - | 3 | 1104 | ||
| AK012122.1 | - | 3 | 1149 | |||
| AK049515.1 | - | 2 | 1121 | |||
| AK090016.1 | - | 3 | 1147 | |||