siRNA Details
| Gene Symbol: E130201H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024324.1 | 1 | 100 | |||
| GenBank mRNA | AK021400.1 | - | 1 | 102 | ||
| BC094539.1 | - | 1 | 75 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130201H02Rik| Gene Symbol: E130201H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024324.1 | 1 | 100 | |||
| GenBank mRNA | AK021400.1 | - | 1 | 102 | ||
| BC094539.1 | - | 1 | 75 | |||