siRNA Details
| Gene Symbol: E130201H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024324.1 | 1 | 702 | |||
| GenBank mRNA | AK021400.1 | - | 1 | 704 | ||
| BC094539.1 | - | 1 | 677 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB542603.1 | - | 1 | 156 | Chr. 7 - 120598176 - 120598472 [+] on Build GRCm38 | |

E130201H02Rik
Genomic Map