siRNA Details
| Gene Symbol: Hnf4aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027970.1 | 4 | 706 | |||
| GenBank mRNA | AK002328.1 | - | 3 | 501 | ||
| BC025862.1 | - | 4 | 696 | 5050923 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnf4aos| Gene Symbol: Hnf4aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027970.1 | 4 | 706 | |||
| GenBank mRNA | AK002328.1 | - | 3 | 501 | ||
| BC025862.1 | - | 4 | 696 | 5050923 | ||