siRNA Details
| Gene Symbol: Hnf4aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027970.1 | 2 | 356 | |||
| GenBank mRNA | BC025862.1 | - | 2 | 346 | 5050923 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnf4aos| Gene Symbol: Hnf4aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027970.1 | 2 | 356 | |||
| GenBank mRNA | BC025862.1 | - | 2 | 346 | 5050923 | |