siRNA Details
Gene Symbol: Hnf4aos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027970.1 | 4 | 809 | |||
GenBank mRNA | AK002328.1 | - | 3 | 604 | ||
BC025862.1 | - | 4 | 799 | 5050923 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hnf4aos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027970.1 | 4 | 809 | |||
GenBank mRNA | AK002328.1 | - | 3 | 604 | ||
BC025862.1 | - | 4 | 799 | 5050923 |