siRNA Details
| Gene Symbol: Speer5-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001582.2 | 7 | 1110 | |||
| NR_027506.1 | 4 | 648 | ||||
| GenBank mRNA | BB679670.1 | - | 1 | 91 | ||
| DV646842.1 | - | 4 | 648 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Speer5-ps1| Gene Symbol: Speer5-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001582.2 | 7 | 1110 | |||
| NR_027506.1 | 4 | 648 | ||||
| GenBank mRNA | BB679670.1 | - | 1 | 91 | ||
| DV646842.1 | - | 4 | 648 | |||