siRNA Details
| Gene Symbol: Speer5-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001582.2 | 6 | 798 | |||
| NR_027506.1 | 3 | 336 | ||||
| GenBank mRNA | DV646842.1 | - | 3 | 336 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Speer5-ps1| Gene Symbol: Speer5-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001582.2 | 6 | 798 | |||
| NR_027506.1 | 3 | 336 | ||||
| GenBank mRNA | DV646842.1 | - | 3 | 336 | ||