siRNA Details
Gene Symbol: Slc22a13b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033303.1 | 6 | 1357 | |||
GenBank mRNA | AK079093.1 | - | 6 | 1284 | ||
BC147282.1 | - | 6 | 1081 | 9056356 | ||
BC147283.1 | - | 6 | 1081 | 9056343 | ||
BY384836.1 | - | 1 | 332 |
