siRNA Details
| Gene Symbol: Slc22a13b-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033303.1 | 6 | 1221 | |||
| GenBank mRNA | AK079093.1 | - | 6 | 1148 | ||
| BC147282.1 | - | 6 | 945 | 9056356 | ||
| BC147283.1 | - | 6 | 945 | 9056343 | ||
| BY384836.1 | - | 1 | 196 | |||

Slc22a13b-ps
Genomic Map