siRNA Details
Gene Symbol: Slc22a13b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033303.1 | 6 | 1442 | |||
GenBank mRNA | AK079093.1 | - | 6 | 1369 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc22a13b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033303.1 | 6 | 1442 | |||
GenBank mRNA | AK079093.1 | - | 6 | 1369 |