siRNA Details
Gene Symbol: Slc22a13b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033303.1 | 2 | 395 | |||
GenBank mRNA | AK079093.1 | - | 2 | 322 | ||
BC147282.1 | - | 2 | 119 | 9056356 | ||
BC147283.1 | - | 2 | 119 | 9056343 |
