siRNA Details
| Gene Symbol: 1700026D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028286.1 | 1 | 68 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF981910.1 | - | 1 | 52 | Chr. 2 - 132490744 - 132512300 [+] on Build GRCm38 | |

1700026D11Rik
Genomic Map