siRNA Details
| Gene Symbol: 1700026D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028286.1 | 4 | 818 | |||
| GenBank mRNA | AK006376.1 | - | 4 | 819 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700026D11Rik| Gene Symbol: 1700026D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028286.1 | 4 | 818 | |||
| GenBank mRNA | AK006376.1 | - | 4 | 819 | ||