siRNA Details
| Gene Symbol: 1700026D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028286.1 | 3 | 422 | |||
| GenBank mRNA | AK006376.1 | - | 3 | 423 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX519837.1 | - | 4 | 311 | Chr. 2 - 132490751 - 132523858 [+] on Build GRCm38 | |

1700026D11Rik
Genomic Map