siRNA Details
| Gene Symbol: B430319G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029474.1 | 1 | 1802 | |||
| GenBank mRNA | AK044362.1 | - | 1 | 1285 | ||
| AK086371.1 | - | 1 | 1709 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B430319G15Rik| Gene Symbol: B430319G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029474.1 | 1 | 1802 | |||
| GenBank mRNA | AK044362.1 | - | 1 | 1285 | ||
| AK086371.1 | - | 1 | 1709 | |||