siRNA Details
| Gene Symbol: 1810032O08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027819.1 | 6 | 779 | |||
| NR_027820.1 | 5 | 587 | ||||
| NR_027821.1 | 5 | 1911 | ||||
| GenBank mRNA | AK007675.1 | - | 6 | 780 | ||
| AK013681.1 | - | 5 | 579 | |||
| AK088915.1 | - | 6 | 780 | |||
| BC055707.1 | - | 5 | 1897 | |||

1810032O08Rik
Genomic Map