siRNA Details
| Gene Symbol: 5430405H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015591.1 | 1 | 233 | |||
| NR_027914.1 | 1 | 233 | ||||
| GenBank mRNA | AK012005.1 | - | 1 | 240 | ||
| AK017271.1 | - | 252 | ||||
| AK134903.1 | - | 1 | 235 | |||
| AK134962.1 | - | 1 | 183 | |||
| BC042697.1 | - | 1 | 54 | |||

5430405H02Rik
Genomic Map