siRNA Details
| Gene Symbol: Pantr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027826.1 | 2 | 126 | |||
| GenBank mRNA | AK011437.1 | - | 2 | 126 | ||
| AK019154.1 | - | 2 | 126 | |||
| BC049671.1 | - | 2 | 131 | |||
| BC058417.1 | - | 2 | 219 | |||
| BC059892.1 | - | 2 | 200 | |||
| BC094545.1 | - | 106 | ||||

Pantr1
Genomic Map