siRNA Details
Gene Symbol: 2010204K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027924.1 | 2 | 525 | |||
NR_027925.1 | 2 | 535 | ||||
GenBank mRNA | AF038507.1 | - | 2 | 489 | ||
AK002502.1 | - | 874 | ||||
AK008437.1 | - | 2 | 526 | |||
AK077356.1 | - | 2 | 569 | |||
BC042200.1 | - | 2 | 488 | |||
BC061056.1 | - | 2 | 536 | |||
BC072610.1 | - | 2 | 449 |
