siRNA Details
| Gene Symbol: 1700018B24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003617.1 | 1 | 185 | |||
| GenBank mRNA | AK006080.1 | - | 1 | 187 | ||
| AK006613.1 | - | 1 | 187 | |||
| AK018885.1 | - | 1 | 156 | |||
| BC049578.1 | - | 1 | 170 | |||
| BC061086.1 | - | 1 | 186 | |||

1700018B24Rik
Genomic Map