siRNA Details
Gene Symbol: 2610203C22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015470.1 | 5 | 628 | |||
GenBank mRNA | BC037765.1 | - | 5 | 628 | ||
BC064105.1 | - | 5 | 628 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2610203C22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015470.1 | 5 | 628 | |||
GenBank mRNA | BC037765.1 | - | 5 | 628 | ||
BC064105.1 | - | 5 | 628 |