siRNA Details
| Gene Symbol: 2610203C22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015470.1 | 5 | 1188 | |||
| GenBank mRNA | BC037765.1 | - | 5 | 1188 | ||
| BC064105.1 | - | 5 | 1188 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610203C22Rik| Gene Symbol: 2610203C22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015470.1 | 5 | 1188 | |||
| GenBank mRNA | BC037765.1 | - | 5 | 1188 | ||
| BC064105.1 | - | 5 | 1188 | |||