siRNA Details
| Gene Symbol: 0610039K10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028113.1 | 1 | 914 | |||
| GenBank mRNA | AK002842.1 | - | 1 | 915 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

0610039K10Rik| Gene Symbol: 0610039K10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028113.1 | 1 | 914 | |||
| GenBank mRNA | AK002842.1 | - | 1 | 915 | ||