siRNA Details
| Gene Symbol: Gm10190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028385.1 | 1 | 1381 | |||
| GenBank mRNA | AK034626.1 | - | 1 | 1382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10190| Gene Symbol: Gm10190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028385.1 | 1 | 1381 | |||
| GenBank mRNA | AK034626.1 | - | 1 | 1382 | ||