siRNA Details
| Gene Symbol: Gm5468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027376.1 | 1 | 915 | |||
| GenBank mRNA | AK139483.1 | - | 1 | 50 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB642134.1 | - | 1 | 44 | Chr. 15 - 25415063 - 25450972 [+] on Build GRCm38 | |

Gm5468
Genomic Map