siRNA Details
| Gene Symbol: Atad3aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027971.1 | 2 | 402 | |||
| NR_027972.1 | 2 | 402 | ||||
| GenBank mRNA | AK011879.1 | - | 2 | 374 | ||
| AK078249.1 | - | 2 | 403 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atad3aos| Gene Symbol: Atad3aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027971.1 | 2 | 402 | |||
| NR_027972.1 | 2 | 402 | ||||
| GenBank mRNA | AK011879.1 | - | 2 | 374 | ||
| AK078249.1 | - | 2 | 403 | |||