siRNA Details
| Gene Symbol: D730005E14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030675.1 | 1 | 1372 | |||
| GenBank mRNA | AK035732.1 | - | 1 | 1374 | ||
| AK037689.1 | - | 1 | 616 | |||
| AK143849.1 | - | 1 | 1357 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D730005E14Rik| Gene Symbol: D730005E14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030675.1 | 1 | 1372 | |||
| GenBank mRNA | AK035732.1 | - | 1 | 1374 | ||
| AK037689.1 | - | 1 | 616 | |||
| AK143849.1 | - | 1 | 1357 | |||