siRNA Details
| Gene Symbol: Gm11978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028586.1 | 6 | 1077 | |||
| GenBank mRNA | AK132798.1 | - | 6 | 1078 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11978| Gene Symbol: Gm11978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028586.1 | 6 | 1077 | |||
| GenBank mRNA | AK132798.1 | - | 6 | 1078 | ||