siRNA Details
| Gene Symbol: Mcpt-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028284.1 | 2 | 123 | |||
| GenBank mRNA | BY328253.1 | - | 2 | 124 | ||
| X78543.1 | - | 2 | 102 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcpt-ps1| Gene Symbol: Mcpt-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028284.1 | 2 | 123 | |||
| GenBank mRNA | BY328253.1 | - | 2 | 124 | ||
| X78543.1 | - | 2 | 102 | |||