siRNA Details
| Gene Symbol: E130112N10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015604.2 | 2 | 1151 | |||
| GenBank mRNA | AK087434.1 | - | 2 | 1152 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB541225.2 | - | 1 | 472 | Chr. 6 - 125239260 - 125239930 [+] on Build GRCm38 | |

E130112N10Rik
Genomic Map