siRNA Details
Gene Symbol: E130112N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015604.2 | 2 | 746 | |||
GenBank mRNA | AK087434.1 | - | 2 | 747 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: E130112N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015604.2 | 2 | 746 | |||
GenBank mRNA | AK087434.1 | - | 2 | 747 |