siRNA Details
| Gene Symbol: E130112N10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015604.2 | 2 | 426 | |||
| GenBank mRNA | AK087434.1 | - | 2 | 427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130112N10Rik| Gene Symbol: E130112N10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015604.2 | 2 | 426 | |||
| GenBank mRNA | AK087434.1 | - | 2 | 427 | ||