siRNA Details
| Gene Symbol: Zbtb11os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027965.1 | 1 | 1235 | |||
| GenBank mRNA | AK006543.1 | - | 1 | 1033 | ||
| AK010004.1 | - | 1 | 1210 | |||
| BC017620.1 | - | 1 | 1169 | 4924337 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_133677.1 | - | 1169 | Chr. - - [-] on Build | ||

Zbtb11os1
Genomic Map